missing translation for 'onlineSavingsMsg'
Learn More

Platinum(IV) nitrate, solution, Pt 15% w/w

Catalog No. AAH3773703
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3773703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3773703 Supplier Thermo Scientific Chemicals Supplier No. H3773703

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 10102-09-7 | N4O12Pt

Platinum(IV) nitrate acts as a precursor in the production of platinum dioxide on support surface.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10102-09-7
Molecular Formula N4O12Pt
MDL Number MFCD25372548
Synonym platinum 4+ ion nitric acid

Specifications

Odor Acidic
Quantity 1 g
UN Number UN3093
Solubility Information Miscible with water.
Formula Weight 443.1
Assay Pt 15% w/w
Chemical Name or Material Platinum(IV) nitrate, solution
Hazard Category H290-H314-H332
Hazard Statement GHS H Statement
H272-H314-H318-H290-H317
May intensify fire; oxidizer.
Causes severe skin burns and eye damage.
Causes serious eye damage.
May be corrosive to metals.
May cause an allergic skin reaction.
Precautionary Statement P234-P260-P264b-P271-P280-P284-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, OXIDIZING, N.O.S.
EINECSNumber 432-400-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only