missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Platinum, 10% on carbon, dry
Description
Platinum, 10% on carbon, is used as a catalyst for the hydrogenation of alkenes, alkynes, nitro groups, ketones and aromatics. It is also used for hydrogenolysis of cyclopropanes. It is involved in the catalyzes oxidation of alcohols.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Platinum, 10% on carbon, is used as a catalyst for the hydrogenation of alkenes, alkynes, nitro groups, ketones and aromatics. It is also used for hydrogenolysis of cyclopropanes. It is involved in the catalyzes oxidation of alcohols.
Solubility
Insoluble in water.
Notes
Store in a cool location. Incompatible with oxidizing agents.
Platinum, 10% on carbon, is used as a catalyst for the hydrogenation of alkenes, alkynes, nitro groups, ketones and aromatics. It is also used for hydrogenolysis of cyclopropanes. It is involved in the catalyzes oxidation of alcohols.
Solubility
Insoluble in water.
Notes
Store in a cool location. Incompatible with oxidizing agents.
Specifications
Specifications
| Quantity | 1 g |
| Odor | Odorless |
| UN Number | UN1325 |
| Merck Index | 14,7529 |
| Solubility Information | Insoluble in water. |
| Chemical Name or Material | Platinum, 10% on carbon, dry |
Safety and Handling
| missing translation for 'healthHazard1' | H228 |
| missing translation for 'healthHazard2' | GHS H Statement H228 Flammable solid. |
| missing translation for 'healthHazard3' | P210-P240-P241-P280-P370+P378q |
| DOTInformation | Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S. |
| EINECSNumber | 231-116-1 |
| RTECSNumber | TP2160000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only