missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Micronova™ Plastic Squeegee
These durably constructed squeegees are made with a polypropylene frame and a choice of either solid rubber or double foam blades for use in wash and production areas. Designed with an ACME threaded handle connection for use with most threaded plastic handles.
$123.28
Spécifications
| Product Type | Squeegee, ACME Threaded End |
|---|---|
| Content And Storage | Store in cool dry area when not in use. |
| Disposable | Reusable |
| For Use With (Application) | Squeegee for Floors, Walls and Windows |
| For Use With (Equipment) | STA Series Plastic Handles |
| Numéro de catalogue | Numéro du manufacturier. | Color | Material | Prix | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Color | Material | Prix | Quantity & Availability | ||||
|
19263618
|
Micronova™
SQDB24R |
Dark Blue | Rubber Blade, Polypropylene Frame |
chaque for $123.28
caisse de 6 chaque for $739.68
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
|||
|
19263619
|
Micronova™
SQDG24R |
Dark Green | Rubber Blade, Polypropylene Frame |
chaque for $123.28
caisse de 6 chaque for $739.68
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
|||
|
19263617
|
Micronova™
SQWH24R |
White | Rubber Blade, Polypropylene Frame |
chaque for $123.28
caisse de 6 chaque for $739.68
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
|||
Description
The Micronova Plastic Squeegees easily clean smooth or uneven surfaces and minimize contamination areas by reducing dirt and bacteria growth. Designed for use with STAH or STAEH series plastic handles. Available in a variety of colors.
- Durably constructed polypropylene squeegees
- Choice of solid rubber or double foam blades
- Excellent for use on smooth surfaces and uneven floor surfaces
- Prevents hazardous sparks when cleaning-up flammable fluids
- Available in 18 and 24 inch (45 and 60 cm) widths
- Use with STAH or STAEH handles for a secure fit
- Will not rust
Spécifications
| Squeegee, ACME Threaded End | |
| Reusable | |
| STA Series Plastic Handles |
| Store in cool dry area when not in use. | |
| Squeegee for Floors, Walls and Windows |