Learn More
Fisherbrand™ Plastic pH Test Strips
Offer excellent color separation in the pH range; 100 pH test strips/vial.
Supplier: Fisherbrand™ 13640519/EMD
Description
- Packaged in flip-top, spill-proof vial
- Colorimetric chart located on the vial label
- Plastic pH test strip construction
- Ideal for lab, field or classroom useInstant,
- Instant dip and read result
Order Info
Box 6 Vials, 100 strips per vial.
Chemical Identifiers
| For lab, field or classroom use, instant dip and read result | |
| Colorimetric chart on the vial label | |
| 100 Strips/Vial, 6 Vials/Pk. | |
| Room Temperature |
Specifications
| 5 to 9 | |
| 0.5 | |
| 100 Strips/Vial, 6 Vials/Pk. | |
| pH Test Strip |
| For lab, field or classroom use, instant dip and read result | |
| Colorimetric chart on the vial label | |
| Room Temperature |