missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Plastic pH Test Strips

Catalog No. p-4604386
Change view
Click to view available options
pH Range:
0 to 6
5 to 9
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. pH Range
S04677 0 to 6
S04673 5 to 9
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S04677 Supplier Fisherbrand™ Supplier No. 13640520/EMD
Only null left

Offer excellent color separation in the pH range; 100 pH test strips/vial.

  • Packaged in flip-top, spill-proof vial
  • Colorimetric chart located on the vial label
  • Plastic pH test strip construction
  • Ideal for lab, field or classroom useInstant,
  • Instant dip and read result

Order Info

Box 6 Vials, 100 strips per vial.

Specifications

pH Range 0 to 6
For Use With (Application) For lab, field or classroom use, instant dip and read result
Graduations 0.5
Includes Colorimetric chart on the vial label
Quantity 100 Strips/Vial, 6 Vials/Pk.
Storage Requirements Room Temperature
Type pH Test Strip