missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Plastic Desiccator

Lightweight and easy to move, this plastic desiccator helps to preserve moisture sensitive items in the classroom or laboratory.

Supplier:  Eisco™ CH0366C

 View more versions of this product

Catalog No. S27045


Only null left

Description

Description

  • Autoclavable
  • Made of polypropylene and polycarbonate
  • Holds vacuum up to 740 mm of Hg for 24 hrs
  • Internal groove is provided on the flange to hold a silicon rubber O-ring
  • Polypropylene stopcock fitted with a self lubricating PTFE plug
  • PTFE Plug works in three ways: vacuum creation, shutting off, and vacuum releasing
Specifications

Chemical Identifiers

Plastic
Vacuum Desiccator

Specifications

Plastic
11 in.
33 cm
10 in.
25.4 cm
1.85 kg
Vacuum Desiccator
27.94 cm
13 in.
20 mm
4.1 lb.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions