missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Plastic Additive 4 Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™
Spécifications
Spécifications
| CAS | 2082-79-3 |
| Purity Grade Notes | These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available. |
| Linear Formula | [(CH3)3 C]2C6H2(OH)CH2 CH2 CO2(CH2)17 CH3 |
| MDL Number | MFCD00017498 |
| Melting Point | 50°C to 52°C |
| Synonym | Octadecyl 3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate |
| Type | Certified Reference Material |
| Quantity | 1 g |
| Formula Weight | 530.86 |
| Grade | Certified Reference Material |