missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Plano Glass Mirrors - Unmounted

Rectangular, plano glass mirrors have a silvered back with protective coating and are designed for physics and educational uses. Pack of 10

$26.32 - $96.21

Specifications

Quantity 10 Pk.
Material Plano Glass
For Use With (Application) Light, Optics
Product Type Mirror
Shape Rectangular
View More Specs

Products 4
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity & Availability  
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity & Availability  
S41514
View Documents Promotion Details
Eisco™
PH0514A
75 x 25 mm (3 x 1 in.)
Each for $26.32
Only null left
 
S41516
View Documents Promotion Details
Eisco™
PH0514D
150 x 25 mm (6 x 1 in.)
Each for $43.64
Only null left
 
S41517
View Documents Promotion Details
Eisco™
PH0514E
150 x 50 mm (6 x 2 in.)
Each for $58.54
Only null left
 
S41518
View Documents Promotion Details
Eisco™
PH0514F
150 x 100 mm (6 x 4 in.)
Each for $96.21
Only null left
 
Description

Description

  • Rectangular shaped plano glass mirrors
  • Designed for educational use
  • Include silvered back with protective coating
  • Pack of 10

Warranty

1 year limited manufacturer warranty.

Specifications

Specifications

10 Pk.
Light, Optics
Rectangular
Plano Glass
Mirror
Videos
SDS
Documents

Documents