missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PLA2G2A Recombinant Rabbit Monoclonal Antibody (23GB2190)
GREENER_CHOICE

Catalog No. MA552482
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA552482 100 μL
MA552483 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA552482 Supplier Invitrogen™ Supplier No. MA552482
Only null left

Rabbit Recombinant Monoclonal Antibody

PLA2G2A Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow

The protein encoded by this gene is a member of the phospholipase A2 family (PLA2). PLA2s constitute a diverse family of enzymes with respect to sequence, function, localization, and divalent cation requirements. This gene product belongs to group II, which contains secreted form of PLA2, an extracellular enzyme that has a low molecular mass and requires calcium ions for catalysis. It catalyzes the hydrolysis of the sn-2 fatty acid acyl ester bond of phosphoglycerides, releasing free fatty acids and lysophospholipids, and thought to participate in the regulation of the phospholipid metabolism in biomembranes. Several alternatively spliced transcript variants with different 5' UTRs have been found for this gene.

Specifications

Antigen PLA2G2A
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB2190
Concentration 0.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene PLA2G2A
Gene Accession No. P14423, P14555, P31482
Gene Alias EC 3.1.1.4; EF; Enhancing factor; eted enzyme type IIA phospholipase A2; GIIC sPLA2; Group IIA phospholipase A2; modifier of Min1; Mom1; non-pancreatic secreted type II phospholipase A2; non-pancreatic secretory phospholipase A2; NPS-PLA2; phosphatidylcholine 2-acylhydrolase 2A; phospholipase A2 group IIA; phospholipase A2, group IIA (platelets, synovial fluid); phospholipase A2, membrane associated; Pla2; PLA2B; PLA2BPLA2; PLA2G2A; PLA2L; PLA2LPLA2S; PLA2S; PLAS1; platelet phospholipase A2; RASF-A; secretory group II phospholipase A2; sPLA2; sPla2-IIA
Gene Symbols PLA2G2A
Host Species Rabbit
Immunogen A synthesized peptide derived from human PLA2G2A (105-144AA).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18780, 29692, 5320
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less