missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PKM2 Recombinant Rabbit Monoclonal Antibody (23GB2920)

Catalog No. MA552533
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA552533 20 μL
MA552532 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA552533 Supplier Invitrogen™ Supplier No. MA552533
Only null left

Rabbit Recombinant Monoclonal Antibody

PKM2 Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot

PKM2 is a pyruvate kinase that catalyzes the production of phosphoenolpyruvate from pyruvate and ATP. PKM2 has been shown to interact with thyroid hormone, and thus may mediate cellular metabolic effects induced by thyroid hormones. PKM2 has been found to bind Opa protein, a bacterial outer membrane protein involved in gonococcal adherence to and invasion of human cells, suggesting a role of this protein in bacterial pathogenesis. Three alternatively spliced transcript variants encoding two distinct isoforms of PKM2 have been reported.

Specifications

Antigen PKM2
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB2920
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene PKM
Gene Accession No. P11980, P14618, P52480
Gene Alias AA414905; AL024370; AL024424; ATP:pyruvate 2-o-phosphotransferase; CTHBP; Cytosolic thyroid hormone-binding protein; epididymis secretory protein Li 30; HEL-S-30; isozymes M1/M2; m2pk; muscle pyruvate kinase; OIP3; OIP-3; OPA-interacting protein 3; p58; PK, muscle type; PK2; Pk-2; Pk3; Pk-3; PKLR; PKM; pkm.L; PKM12; PKM2; Pykm; pyruvate kinase; pyruvate kinase 2/3; pyruvate kinase 3; pyruvate kinase isozymes M1/M2; pyruvate kinase M1/2; pyruvate kinase M1/2 L homeolog; pyruvate kinase M1/2 L homeolog; pyruvate kinase, muscle; pyruvate kinase muscle isozyme; Pyruvate kinase PKM; pyruvate kinase, liver and RBC; pyruvate kinase, muscle; TCB; THBP1; thyroid hormone-binding protein 1; thyroid hormone-binding protein, cytosolic; tu; tumor M2-PK; XELAEV_18018020mg
Gene Symbols PKM
Host Species Rabbit
Immunogen A synthesized peptide derived from human PKM2 (50-100AA).
Purification Method Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18746, 25630, 5315
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less