missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PKC mu Recombinant Rabbit Monoclonal Antibody (23GB4590)
GREENER_CHOICE

Catalog No. MA552710
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA552710 100 μL
MA552711 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA552710 Supplier Invitrogen™ Supplier No. MA552710
Only null left

Rabbit Recombinant Monoclonal Antibody

PKC mu Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow

The PKC family of serine/threonine kinases, including PRKCN (PKD3), is activated intracellularly by signal transduction pathways. In humans, at least 12 different PKC polypeptides have been identified. These isoforms differ in primary structure, tissue distribution, subcellular localization, mode of action in vitro, response to extracellular signals, and substrate specificity. PKC alpha, beta I, beta II and gamma form the conventional family; their activities are Ca2+- and phospholipid-dependent. PKC mu activation is dependent on the phosphorylation of two activation loop sites, Ser744 and Ser748, via a PKC-dependent signaling pathway.

Specifications

Antigen PKC mu
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB4590
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Prkd3
Gene Accession No. O94806, Q15139, Q5XIS9, Q62101, Q8BZ03, Q8K1Y2, Q9BZL6, Q9WTQ1
Gene Alias 4930557O20Rik; 5730497N19Rik; AI325941; DKFZP586E0820; EPK2; HSPC187; nPKC-D1; nPKC-D2; nPKC-mu; nPKC-nu; PKC mu; PKC nu; PKC u; PKC v; Pkcm; PKCmu; PKC-MU; Pkcnu; PKC-NU; PKCu; PKC-u; PKCv; PKC-v; Pkd; PKD1; PKD2; PKD3; Prkcm; PRKCN; Prkd1; PRKD2; PRKD3; Protein kinase C mu type; protein kinase C nu type; protein kinase C, mu; protein kinase C, nu; protein kinase D; protein kinase D1; protein kinase D2; protein kinase D3; protein kinase EPK2; RGD1308054; Serine/threonine-protein kinase D1; serine/threonine-protein kinase D2; Serine/threonine-protein kinase D3
Gene Symbols PRKD1, Prkd2, Prkd3
Host Species Rabbit
Immunogen A synthesized peptide derived from human PKC mu (200-266AA).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 101540, 18760, 23683, 25865, 292658, 313834, 5587, 75292, 85421
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less