missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PKC delta/theta Recombinant Rabbit Monoclonal Antibody (23GB4585)

Catalog No. MA553775
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA553775 20 μL
MA553774 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA553775 Supplier Invitrogen™ Supplier No. MA553775
Only null left

Rabbit Recombinant Monoclonal Antibody

PKC delta/theta Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot

PRKCD (Protein Kinase C Delta) is a Protein Coding gene. Diseases associated with PRKCD includeAutoimmune Lymphoproliferative Syndrome, Type IiiandSystemic Lupus Erythematosus 16. Among its related pathways areRole of Calcineurin dependent NFAT signaling in lymphocytesandWnt Signaling Pathway and Pluripotency. Gene Ontology (GO) annotations related to this gene includetransferase activity, transferring phosphorus containing groupsandprotein tyrosine kinase activity. An important paralog of this gene isPRKCQ (PKC theta).

Specifications

Antigen PKC delta/theta
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB4585
Concentration 3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene PRKCQ
Gene Accession No. P09215, P28867, Q02111, Q04759, Q05655, Q9WTQ0
Gene Alias A130035A12Rik; AI385711; ALPS3; AW494342; CVID9; D14Ertd420e; may1; npkc-delta; nPKC-theta; PKC theta; PKC[d]; PKC-0; Pkcd; PKCd1; PKCd2; PKCdelta; PKC-delta; PKCdelta1; pkc-delta1; PKCdelta2; Pkcq; PKCtheta; PKC-theta; Prkcd; prkcd.L; prkcd.S; prkcd-a; prkcd-b; PRKCQ; PRKCT; protein kinase C delta; protein kinase C delta S homeolog; protein kinase C delta type; Protein kinase C delta type catalytic subunit; Protein kinase C delta type regulatory subunit; protein kinase C delta variant IX; protein kinase C delta VIII; protein kinase C theta; protein kinase C theta type; protein kinase C, delta; protein kinase C, delta IV; protein kinase C, delta L homeolog; protein kinase C, delta V; protein kinase C, theta; protein kinase C[d]; protein kinase delta1; SDK1; SDK-1; Sphingosine-dependent protein kinase-1; Tyrosine-protein kinase PRKCD; XELAEV_18025733mg
Gene Symbols PRKCD, PRKCQ
Host Species Rabbit
Immunogen A synthesized peptide derived from human PKC delta+PKC theta (100-350AA).
Purification Method Affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 170538, 18753, 18761, 5580, 5588, 85420
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less