missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PKC delta Recombinant Rabbit Monoclonal Antibody (23GB2410)
GREENER_CHOICE

Catalog No. MA553546
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA553546 100 μL
MA553547 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA553546 Supplier Invitrogen™ Supplier No. MA553546
Only null left

Rabbit Recombinant Monoclonal Antibody

PKC delta Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot

The PKC family of serine/threonine kinases, including PRKCD (PKC delta), is activated intracellularly by signal transduction pathways. In humans, at least 12 different PKC polypeptides have been identified. These isoforms differ in primary structure, tissue distribution, subcellular localization, mode of action in vitro, response to extracellular signals, and substrate specificity. PKC alpha, beta I, beta II, and gamma form the conventional family; their activities are Ca2+- and phospholipid-dependent. Protein kinase C (PKC) can be activated by calcium and the second messenger diacylglycerol. PKC family members phosphorylate a wide variety of protein targets and are known to be involved in diverse cellular signaling pathways. PKC family members also serve as major receptors for phorbol esters, a class of tumor promoters. Each member of the PKC family has a specific expression profile and is believed to play distinct roles in cells. The protein encoded by this gene is one of the PKC family members. Studies both in human and mice demonstrate that this kinase is involved in B cell signaling and in the regulation of growth, apoptosis, and differentiation of a variety of cell types. Alternatively spliced transcript variants encoding the same protein have been observed.

Specifications

Antigen PKC delta
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB2410
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene PRKCD
Gene Accession No. P28867, Q05655
Gene Alias 1-May; AI385711; ALPS3; CVID9; D14Ertd420e; kinase nPKC-delta; KPCD; MAY1; nPKC-delta; PKC[d]; Pkcd; PKCdelta; Prkcd; protein kinase C delta; protein kinase C delta type; Protein kinase C delta type catalytic subunit; Protein kinase C delta type regulatory subunit; protein kinase C delta variant IX; protein kinase C delta VIII; protein kinase C, delta; protein kinase C, delta IV; protein kinase C, delta V; protein kinase C[d]; SDK1; Sphingosine-dependent protein kinase-1; Tyrosine-protein kinase PRKCD
Gene Symbols PRKCD
Host Species Rabbit
Immunogen A synthesized peptide derived from human PKC delta (500-676AA).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18753, 5580
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less