missing translation for 'onlineSavingsMsg'
Learn More

pJTI™ R4 CMV-TO MCS pA Vector

Catalog No. A15004
Encompass
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A15004 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A15004 Supplier Thermo Fisher Scientific Supplier No. A15004

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

pJTI™ R4 CMV-TO MCS pA Vector

The pJTI™ R4 CMV-TO MCS pA vector is designed for the expression of your gene of interest under the control of the tet-inducible CMV promoter after restriction enzyme cloning and retargeting into the genomic R4 site of a Jump-In™ parental cell line.

This vector can be used for inducible or constitutive expression of your gene of interest, depending on which Jump-In™ parental cell line you use. When this vector is retargeted into a Jump-In™ T-REx™ parental cell line, gene expression is controlled by the tet-operon and can be induced by adding doxycycline to the growth media. When used with a Jump-In™ parental cell line such as the Jump-In™ GripTite™ HEK293 cell line, constitutive expression is achieved after retargeting. The R4 sites in the Jump-In™ parental cell lines result in a high retargeting efficiency, requiring less effort and less time than traditional cell engineering methods. Retargeting of Jump-In™ parental cell lines results in creation of an isogenic pool that is sufficient for cell-based experiments without the need for clonal selection. Alternatively, the high retargeting efficiency allows for easy selection of a positive stable clone for expressing your gene of interest.

Specifications

Constitutive or Inducible System Inducible
Delivery Type Transfection
Inducing Agent Tetracycline
Promoter CMV/TO
Product Type Mammalian Expression Vector
Selection Agent (Eukaryotic) Geneticin™ (G-418)
Content And Storage pJTI™ R4 CMV-TO MCS pA vector (100 μg), store at -20 °C
Antibiotic Resistance Bacterial Ampicillin (AmpR)
Protein Tag Untagged
Cloning Method Restriction Enzyme/MCS
Quantity 100 μg
Vector Jump-In Vectors
Product Line Jump-In
For Use With (Application) Stable Cell Line Development, Targeted Integration, Regulated Expression
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.