missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pizotifen 98.0+%, TCI America™
$182.68 - $182.68
Chemical Identifiers
| CAS | 15574-96-6 |
|---|---|
| Molecular Formula | C19H21NS |
| Molecular Weight (g/mol) | 295.444 |
| MDL Number | MFCD00864192 |
| InChI Key | FIADGNVRKBPQEU-UHFFFAOYSA-N |
| Synonym | 4-[9,10-Dihydro-4H-benzo[4,5]cyclohepta[1,2-b]thien-4-ylidene]-1-methylpiperidine, Sandomigran |
| PubChem CID | 27400 |
| ChEBI | CHEBI:50212 |
| IUPAC Name | 4-(4,5-dihydrobenzo[1,2]cyclohepta[3,4-b]thiophen-10-ylidene)-1-methylpiperidine |
| SMILES | CN1CCC(=C2C3=C(CCC4=CC=CC=C42)SC=C3)CC1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
P2344200MG
|
TCI America
P2344200MG |
200 mg |
Each for $182.68
|
|
|||||
Chemical Identifiers
| 15574-96-6 | |
| 295.444 | |
| FIADGNVRKBPQEU-UHFFFAOYSA-N | |
| 27400 | |
| 4-(4,5-dihydrobenzo[1,2]cyclohepta[3,4-b]thiophen-10-ylidene)-1-methylpiperidine |
| C19H21NS | |
| MFCD00864192 | |
| 4-[9,10-Dihydro-4H-benzo[4,5]cyclohepta[1,2-b]thien-4-ylidene]-1-methylpiperidine, Sandomigran | |
| CHEBI:50212 | |
| CN1CCC(=C2C3=C(CCC4=CC=CC=C42)SC=C3)CC1 |
Specifications
| 15574-96-6 | |
| White | |
| MFCD00864192 | |
| 4-[9,10-Dihydro-4H-benzo[4,5]cyclohepta[1,2-b]thien-4-ylidene]-1-methylpiperidine, Sandomigran | |
| CN1CCC(=C2C3=C(CCC4=CC=CC=C42)SC=C3)CC1 | |
| 295.444 | |
| CHEBI:50212 | |
| ≥98.0% (HPLC) | |
| Pizotifen |
| 149°C | |
| C19H21NS | |
| 200 mg | |
| FIADGNVRKBPQEU-UHFFFAOYSA-N | |
| 4-(4,5-dihydrobenzo[1,2]cyclohepta[3,4-b]thiophen-10-ylidene)-1-methylpiperidine | |
| 27400 | |
| 295.44 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : TM7165000
TSCA : No