accessibility menu, dialog, popup

UserName

CAS RN 15574-96-6

IUPAC Name: 1-methyl-4-{6-thiatricyclo[8.4.0.0³,⁷]tetradeca-1(14),3(7),4,10,12-pentaen-2-ylidene}piperidine
Synonyms: litec polomigran pizotyline sandomigran pizotifenum 苯噻啶 pizotifen sandomygran pizotifen (product) pizotifeno
Molecular Weight (g/mol): 295.44
Molecular Formula: C19H21NS
InChi Key: FIADGNVRKBPQEU-UHFFFAOYSA-N
SMILES: CN1CCC(CC1)=C1C2=C(CCC3=CC=CC=C13)SC=C2
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results