missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pivalonitrile 98.0+%, TCI America™
Supplier: TCI America P0463100ML
Specifications
| Pivalonitrile | |
| Colorless | |
| C5H9N | |
| 100 mL | |
| pivalonitrile, trimethylacetonitrile, 2,2-dimethylpropionitrile, tert-butyl cyanide, propanenitrile, 2,2-dimethyl, tert-butylnitrile, 2-cyano-2-methylpropane, trimethyl acetonitrile, tert-cyanobutane, t-bucn | |
| CC(C)(C)C#N | |
| 83.134 | |
| 83.13 | |
| Liquid |
| 630-18-2 | |
| 105°C | |
| MFCD00001847 | |
| 1993 | |
| JAMNHZBIQDNHMM-UHFFFAOYSA-N | |
| 2,2-dimethylpropanenitrile | |
| 12416 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 630-18-2 | |
| 83.134 | |
| JAMNHZBIQDNHMM-UHFFFAOYSA-N | |
| 12416 | |
| CC(C)(C)C#N |
| C5H9N | |
| MFCD00001847 | |
| pivalonitrile, trimethylacetonitrile, 2,2-dimethylpropionitrile, tert-butyl cyanide, propanenitrile, 2,2-dimethyl, tert-butylnitrile, 2-cyano-2-methylpropane, trimethyl acetonitrile, tert-cyanobutane, t-bucn | |
| 2,2-dimethylpropanenitrile |
Safety and Handling
TSCA : Yes