missing translation for 'onlineSavingsMsg'
Learn More

Pivalonitrile 98.0+%, TCI America™

Catalog No. p-7134622
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Catalog No. Quantity
P046325ML 25 mL
P0463100ML 100 mL
P0463500ML 500 mL
3 options

Catalog No. P046325ML

Supplier: TCI America P046325ML

Only null left

Chemical Identifiers

CAS 630-18-2
Molecular Formula C5H9N
Molecular Weight (g/mol) 83.134
MDL Number MFCD00001847
InChI Key JAMNHZBIQDNHMM-UHFFFAOYSA-N
Synonym pivalonitrile, trimethylacetonitrile, 2,2-dimethylpropionitrile, tert-butyl cyanide, propanenitrile, 2,2-dimethyl, tert-butylnitrile, 2-cyano-2-methylpropane, trimethyl acetonitrile, tert-cyanobutane, t-bucn
PubChem CID 12416
IUPAC Name 2,2-dimethylpropanenitrile
SMILES CC(C)(C)C#N

Specifications

Color Colorless
Boiling Point 105°C
Quantity 25 mL
UN Number 1993
Formula Weight 83.13
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Pivalonitrile