missing translation for 'onlineSavingsMsg'
Learn More

Pivalaldehyde 95.0+%, TCI America™

Catalog No. P08475ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P08475ML 5 mL
P084725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P08475ML Supplier TCI America Supplier No. P08475ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 630-19-3
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.13
MDL Number MFCD00006962
InChI Key FJJYHTVHBVXEEQ-UHFFFAOYSA-N
Synonym trimethylacetaldehyde, pivalaldehyde, pivaldehyde, propanal, 2,2-dimethyl, neopentanal, pivalic aldehyde, 2,2-dimethylpropionaldehyde, unii-ssc20260qw, trimethyl acetaldehyde, trimethylacetaldehyde pivaldehyde
PubChem CID 12417
IUPAC Name 2,2-dimethylpropanal
SMILES CC(C)(C)C=O

Specifications

Melting Point 6°C
Color Yellow
Boiling Point 75°C
Quantity 5 mL
UN Number 1989
Formula Weight 86.13
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Pivalaldehyde
EINECSNumber (2)-0494
TSCA Yes
Recommended Storage Refrigerator