missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PIST Recombinant Rabbit Monoclonal Antibody (23GB5870)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA553933
Description
PIST Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot
PDZ domains contain approximately 90 amino acids and bind the extreme C terminus of proteins in a sequence-specific manner. PIST, a PDZ domain-containing Golgi protein, was discovered in a yeast two-hybrid system as a binding partner to Beclin-1, a Bcl-2-interacting protein homologous to the yeast autophagy gene apg6. Experiments with mutant PIST proteins lacking the PDZ domain showed that PIST interaction with Beclin-1 through its coiled-coil domain can modulate Beclin-1 activity and suggest that PIST interactions with other proteins through its PDZ domain may regulate the activity of PIST and Beclin-1.Chemical Identifiers
| PIST | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human, Mouse, Rat | |
| GOPC | |
| 2210402P09Rik; AI844555; CAL; CFTR-associated ligand; dJ94G16.2; dJ94G16.2 PIST; FIG; Fused in glioblastoma; golgi associated PDZ and coiled-coil motif containing; Golgi associated PDZ and coiled-coil motif containing protein; golgi-associated PDZ and coiled-coil motif containing protein; Golgi-associated PDZ and coiled-coil motif-containing protein; Gopc; GOPC1; PDZ domain-containing protein PIST; PDZ protein interacting specifically with TC10; PDZ/coiled-coil domain binding partner for the rho-family GTPase TC10; PIST | |
| GOPC | |
| Primary | |
| Affinity chromatography |
| 23GB5870 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8BH60, Q9HD26 | |
| 309774, 57120, 94221 | |
| A synthesized peptide derived from human PIST (1-50AA). | |
| Antibody |
Specifications
| PIST | |
| Recombinant Monoclonal | |
| Unconjugated | |
| GOPC | |
| 2210402P09Rik; AI844555; CAL; CFTR-associated ligand; dJ94G16.2; dJ94G16.2 PIST; FIG; Fused in glioblastoma; golgi associated PDZ and coiled-coil motif containing; Golgi associated PDZ and coiled-coil motif containing protein; golgi-associated PDZ and coiled-coil motif containing protein; Golgi-associated PDZ and coiled-coil motif-containing protein; Gopc; GOPC1; PDZ domain-containing protein PIST; PDZ protein interacting specifically with TC10; PDZ/coiled-coil domain binding partner for the rho-family GTPase TC10; PIST | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 309774, 57120, 94221 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB5870 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8BH60, Q9HD26 | |
| GOPC | |
| A synthesized peptide derived from human PIST (1-50AA). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |