missing translation for 'onlineSavingsMsg'
Learn More

PISD Monoclonal Antibody (OTI4G5)
GREENER_CHOICE

Catalog No. PIMA526861
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA526861 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA526861 Supplier Thermo Fisher Scientific Supplier No. MA526861

PISD Monoclonal Antibody (OTI4G5)

PISD Monoclonal Antibody for Western Blot, IHC (P)

Specifications

Antigen PISD
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI4G5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PISD
Gene Accession No. Q9UG56
Gene Alias 9030221M09Rik; DJ858B16; dJ858B16.2; DKFZp566G2246; phosphatidylserine decarboxylase; Phosphatidylserine decarboxylase alpha chain; Phosphatidylserine decarboxylase beta chain; phosphatidylserine decarboxylase proenzyme; phosphatidylserine decarboxylase proenzyme, mitochondrial; phosphatidylserine decarboxylase proenzyme; phosphatidylserine decarboxylase proenzyme, mitochondrial; Pisd; PSD; PSDC; PSSC
Gene Symbols PISD
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 80-340 of PISD produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23761
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less