missing translation for 'onlineSavingsMsg'
Learn More

Piroxicam

Catalog No. AAJ6323903
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6323903 1 g
AAJ6323906 5 g
AAJ6323909 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6323903 Supplier Thermo Scientific Chemicals Supplier No. J6323903

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 36322-90-4 | C15H13N3O4S | 331.346 g/mol

Piroxicam is an effective and potent inhibitor of prostaglandin synthesis and a Cox-1 and Cox-2 inhibitor. Studies show that Piroxicam blocks release of platelet ADP, and inhibits Cox-1 more potently than Cox-2.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Piroxicam is an effective and potent inhibitor of prostaglandin synthesis and a Cox-1 and Cox-2 inhibitor. Studies show that Piroxicam blocks release of platelet ADP, and inhibits Cox-1 more potently than Cox-2.

Solubility
Soluble in water, ethanol, chloroform, ethyl acetate.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 36322-90-4
Molecular Formula C15H13N3O4S
Molecular Weight (g/mol) 331.346
MDL Number MFCD00057317
InChI Key QYSPLQLAKJAUJT-UHFFFAOYSA-N
Synonym piroxicam, feldene, piroxicamum, pyroxycam, roxicam, piroftal, baxo, artroxicam, bruxicam, caliment
PubChem CID 54676228
ChEBI CHEBI:8249
IUPAC Name 4-hydroxy-2-methyl-1,1-dioxo-N-pyridin-2-yl-1$l^{6},2-benzothiazine-3-carboxamide
SMILES CN1C(=C(C2=CC=CC=C2S1(=O)=O)O)C(=O)NC3=CC=CC=N3

Specifications

Color Yellow
Quantity 1 g
UN Number UN2811
Merck Index 14,7506
Solubility Information Soluble in water,ethanol,chloroform,ethyl acetate.
Formula Weight 331.35
Physical Form Powder
Chemical Name or Material Piroxicam
Hazard Category H301
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P264b-P270-P301+P310-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 252-974-3
RTECSNumber DL0705000
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only