missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Piroctone Olamine Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 68890-66-4 |
| Flash Point | 161.9°C |
| Purity Grade Notes | These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available. |
| Linear Formula | C14H23 NO2 · C2H7 NO |
| MDL Number | MFCD01690792 |
| Synonym | 1-Hydroxy-4-methyl-6-(2,4,4-trimethylpentyl)-2(1 H)-pyridone ethanolammonium salt |
| Type | Certified Reference Material |
| Quantity | 500 mg |
| Formula Weight | 298.42 |
| Grade | Certified Reference Material |