missing translation for 'onlineSavingsMsg'
Learn More

Pirfenidone 98.0+%, TCI America™

Catalog No. p-7135299
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P1871100MG 100 mg
P18711G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P1871100MG Supplier TCI America Supplier No. P1871100MG
Only null left

Chemical Identifiers

CAS 53179-13-8
Molecular Formula C12H11NO
Molecular Weight (g/mol) 185.226
MDL Number MFCD00866047
InChI Key ISWRGOKTTBVCFA-UHFFFAOYSA-N
Synonym pirfenidone, 5-methyl-1-phenylpyridin-2 1h-one, esbriet, deskar, pirespa, amr-69, 5-methyl-1-phenyl-2-1h-pyridone, pirfenidonum, pirfenidona, amr 69
PubChem CID 40632
ChEBI CHEBI:32016
IUPAC Name 5-methyl-1-phenylpyridin-2-one
SMILES CC1=CN(C(=O)C=C1)C2=CC=CC=C2

Specifications

Melting Point 109°C
Quantity 100 mg
Formula Weight 185.23
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Pirfenidone
RTECSNumber UV1148200
TSCA No