accessibility menu, dialog, popup

UserName

CH3NO

CAS RN 53179-13-8

IUPAC Name: 5-methyl-1-phenyl-1,2-dihydropyridin-2-one
Synonyms: pirfenidone pirfenidonum pirfenidona deskar 吡非尼酮
Molecular Weight (g/mol): 185.23
Molecular Formula: C12H11NO
InChi Key: ISWRGOKTTBVCFA-UHFFFAOYSA-N
SMILES: CC1=CN(C(=O)C=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Grade 
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)