missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Pirfenidone
Chemical Identifiers
| CAS | 53179-13-8 |
|---|---|
| Molecular Formula | C12H11NO |
| Molecular Weight (g/mol) | 185.23 |
| InChI Key | ISWRGOKTTBVCFA-UHFFFAOYSA-N |
| IUPAC Name | 5-methyl-1-phenyl-1,2-dihydropyridin-2-one |
| SMILES | CC1=CN(C(=O)C=C1)C1=CC=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC464720010
|
Thermo Scientific™
464720010 |
1 g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC464722500
|
Thermo Scientific™
464722500 |
250 mg |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 53179-13-8 | |
| 185.23 | |
| 5-methyl-1-phenyl-1,2-dihydropyridin-2-one |
| C12H11NO | |
| ISWRGOKTTBVCFA-UHFFFAOYSA-N | |
| CC1=CN(C(=O)C=C1)C1=CC=CC=C1 |
Specifications
| 53179-13-8 | |
| 1 g | |
| ISWRGOKTTBVCFA-UHFFFAOYSA-N | |
| 5-methyl-1-phenyl-1,2-dihydropyridin-2-one | |
| 185.23 |
| C12H11NO | |
| Solubility aq. soln.: insoluble. | |
| CC1=CN(C(=O)C=C1)C1=CC=CC=C1 | |
| 185.23 | |
| Pirfenidone |
Safety and Handling
GHS H Statement Harmful if swallowed.
GHS P Statement IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash face, hands and any exposed skin thoroughly after handling.
GHS Signal Word: Warning
RUO â Research Use Only