missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Pirfenidone

Catalog No. AC464722500
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC464722500 250 mg
AC464720010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC464722500 Supplier Thermo Scientific Chemicals Supplier No. 464722500

CAS: 53179-13-8 | C12H11NO | 185.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 53179-13-8
Molecular Formula C12H11NO
Molecular Weight (g/mol) 185.23
InChI Key ISWRGOKTTBVCFA-UHFFFAOYSA-N
IUPAC Name 5-methyl-1-phenyl-1,2-dihydropyridin-2-one
SMILES CC1=CN(C(=O)C=C1)C1=CC=CC=C1

Specifications

Melting Point 109.0°C
Quantity 250 mg
Solubility Information Solubility aq. soln.: insoluble.
Formula Weight 185.23
Chemical Name or Material Pirfenidone
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Harmful if swallowed.

Precautionary Statement GHS P Statement IF SWALLOWED: rinse mouth.

Do NOT induce vomiting.

Call a POISON CENTER or doctor/physician if you feel unwell.

Wash face, hands and any exposed skin thoroughly after handling.

RUO – Research Use Only