Learn More
Invitrogen™ PIR-A/B Monoclonal Antibody (10-1-PIR), APC, eBioscience™, Invitrogen™

Description
Description: This 10-1-PIR monoclonal antibody reacts with mouse paired Ig-like receptors of activating (PIR)-A and -B. These cell surface glycoproteins, which contain six extracellular Ig-like domains with distinct transmembrane and cytoplasmic regions, are expressed together on B lymphocytes, dendritic cells, macrophages, granulocytes, platelets, and mast cells. However, expression of PIR-A/B has not been observed on T cells, NK cells, and erythrocytes. The PIR-A receptor interacts with signaling molecules containing immunoreceptor tyrosine-based activation motifs (ITAMs) which lead to its activating function. In contrast, the PIR-B receptor associates with proteins possessing immunoreceptor tyrosine-based inhibitory motifs (ITIMs); therefore, this receptor has been shown to have an inhibitory function. PIRs bind MHC class I to modulate cell signaling and homeostasis of the immune system. Moreover, PIR-B knockout mice have been shown to exhibit susceptibility to Salmonella infection. Crossblocking studies indicate that 10-1-PIR recognizes a different epitope from 6C1 (cat. no. 46-5978). Applications Reported: This 10-1-PIR antibody has been reported for use in flow cytometric analysis. Applications Tested: This 10-1-PIR antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.25 μg per test.
Specifications
Specifications
| Antigen | PIR-A/B |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 10-1-PIR |
| Concentration | 0.2 mg/mL |
| Conjugate | APC |
| Formulation | PBS with 0.09% sodium azide; pH 7.2 |
| Gene | Pirb |
| Gene Accession No. | P97484 |
| Gene Alias | 6M21; CD85k; cell-surface glycoprotein p91; Gp49b; Gp91; leukocyte immunoglobulin-like receptor 3; leukocyte immunoglobulin-like receptor subfamily B member 3; leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), member 3; Lilrb3; Lilrb4; LIR-3; Ly89; paired Ig-like receptor B; paired immunoglobulin-like receptor; paired immunoglobulin-like receptor B; paired-Ig-like receptor A1; Pir; Pira1; PIR-A1; Pirb; PIR-B |
| Show More |
For Research Use Only.