missing translation for 'onlineSavingsMsg'
Learn More

Gilson™ PIPETMANTM L Multichannel Pipettes Trade-In Promo
GREENER_CHOICE

Catalog No. FA10039TG
Change view
Click to view available options
Number of Channels:
12
8
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Number of Channels
FA10039TG 8
FA10040TG 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. FA10039TG Supplier Gilson™ Supplier No. FA10039T

Send in used or broken pipettes and save on select Gilson models. All used brands and models qualify and a pre-postage bag will be included with the order. Refurbished pipettes will be donated to Seeding Labs, who distributes to labs and universities around the world who need it most.

Optimized Comfort

  • One of the most comfortable models in the Gilson portfolio
  • Ideal weight and handle shape
  • Natural fit of the pipette allows for a relaxed grip to make pipetting less fatiguing
  • Features an updated piston assembly with more responsive springs to reduce pipetting forces and decrease the risk of repetitive strain injuries
  • Adjustable tip ejector for right- or left-handedness, whichever is most comfortable

Optimized Safety and Precision

  • Volume-locking ensures no accidental volume change during pipetting cycles, which could alter the accuracy and precision of results
  • Patented locking system fully disconnects the operating rod from the counter assembly during pipetting
  • Once locked into place, the volume cannot be accidentally changed to prevent accidental volume drift during pipetting tasks
  • Variable volume models are fully autoclavable to minimize cross-contamination
  • No disassembly required for maximum convenience

Optimized Traceability

  • Features a 2D identification bar code for easy traceability
  • Particularly suited for enhanced GLP compliance
  • Color-coded push button corresponds in color to Gilson PIPETMANTM Tip packages
  • Using a universal scanner download the product part number, date of manufacturing, and serial number

Specifications

Compatible Tips PIPETMAN DIAMOND Tips D1200, DF1200ST
Adjustable Tip Spacing No
Autoclavable Autoclavable
Color Blue
Dispensing Speed Manual
Disposable Reusable
For Use With Pipette Tip 500 to 5000 μL
Grip Type Ergonomic
Material Polypropylene, PVDF, PBT
Model Pipetman™ L
Number of Channels 8
Product Line Pipetman
Programmability No
Sterility Non-sterile
Type Manual Air Displacement Pipette
Volume (Metric) 100 to 1200 μL
Quantity 1
System Type Metal Ejector
Width (English) 5.98 in
Show More Show Less