missing translation for 'onlineSavingsMsg'
Learn More

Gilson™ PIPETMANTM L Multichannel Pipette Trade-In PROMO
GREENER_CHOICE

Catalog No. FA10016TG
Change view
Click to view available options
Color:
Green
Red
White
Yellow
Volume (Metric):
0.5 to 10 μL
2 to 20 μL
20 to 200 μL
20 to 300 μL
Number of Channels:
12
8
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Volume (Metric) Number of Channels
FA10016TG Green 20 to 300 μL 12
FA10015TG Green 20 to 300 μL 8
FA10013TG Red 0.5 to 10 μL 8
FA10014TG Red 0.5 to 10 μL 12
FA10011TG Yellow 20 to 200 μL 8
FA10012TG Yellow 20 to 200 μL 12
FA10009TG White 2 to 20 μL 8
FA10010TG White 2 to 20 μL 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. FA10016TG Supplier Gilson™ Supplier No. FA10016T

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Send in your used or broken pipettes and save on select Gilson models. All used brands and models qualify and a pre-postage bag will be included with the order. Refurbished pipettes will be donated to Seeding Labs, who distributes to labs and universities around the world who need it most.

  • Gilson-patented volume locking system guarantees no accidental change in volume while pipetting
  • Adjustable tip ejector provides comfort for right- and left-handed users
  • Redesigned handle fits naturally in your hand, regardless of the size
  • Unique internal design applies even, equal pressure from either side of the pipettes, rather than on the center of the channels like many other multichannel pipettes do
  • 2D identification code tracks product reference number, date of manufacture, serial number, and maximum volume of pipette
  • Available in 8- or 12- channels, in volumes ranging from 0.50 to 300 μL

Specifications

Compatible Tips PIPETMAN DIAMOND Tips D200, D300, DF200ST, DF300ST
Adjustable Tip Spacing No
Autoclavable Autoclavable
Color Green
Dispensing Speed Manual
Disposable Reusable
For Use With Pipette Tip 50 to 1000 μL
Grip Type Ergonomic
Material Polypropylene
Model P12x300L
Number of Channels 12
Product Line Pipetman
Programmability No
Sterility Non-sterile
Type Pipette
Volume (Metric) 20 to 300 μL
Finger Style Ergonomic
Quantity 1
Width (English) 5.98 in
Fit for Purpose Certified Quality
Show More Show Less