missing translation for 'onlineSavingsMsg'
Learn More

Gilson™ PIPETMANTM L Mechanical Pipette Trade-In PROMO
GREENER_CHOICE

Catalog No. FA10001MTG
Change view
Click to view available options
Volume (Metric):
0.2 to 2 μL
1 to 10 μL
1 to 10 mL
10 to 100 μL
100 to 1000 μL
2 to 20 μL
20 to 200 μL
500 to 5000 μL
Color:
Blue
Orange
Purple
Red
Salmon
Yellow
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Color
FA10001MTG 0.2 to 2 μL Orange
FA10002MTG 1 to 10 μL Red
FA10003MTG 2 to 20 μL Yellow
FA10004MTG 10 to 100 μL Salmon
FA10005MTG 20 to 200 μL Yellow
FA10006MTG 100 to 1000 μL Blue
FA10007TG 500 to 5000 μL Purple
FA10008TG 1 to 10 mL Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. FA10001MTG Supplier Gilson™ Supplier No. FA10001MT

Send in your used or broken pipettes and save on select Gilson models. All used brands and models qualify and a pre-postage bag will be included with the order. Refurbished pipettes will be donated to Seeding Labs, who distributes to labs and universities around the world who need it most.

  • Redesigned handle fits naturally in user's hand, regardless of size
  • Volume locking system guarantees no accidental changes in volume while pipetting
  • Adjustable tip ejector provides comfort for right- and left-handed users
  • 2D identification code allows for easy tracking of product's information
  • New piston assembly designed to drastically improve pipetting and purge forces
  • Redesigned tip ejector shape and internal mechanism reduces tip ejection forces up to 50%

Specifications

Accuracy ±0.024, ±0.025, ±0.027, ±0.03 μL
Compatible Tips Stainless Steel Ejector
Adjustable Tip Spacing No
Autoclavable Autoclavable
Color Orange
Dispensing Speed Manual
Disposable Reusable
For Use With D-10, DL10, DF10ST, DFL10ST Tip
Grip Type Ergonomic
Material Polypropylene
Model P2L
Number of Channels 1
Product Line Pipetman
Programmability No
Sterility Non-sterile
Type Mechanical Pipette
Volume (Metric) 0.2 to 2 μL
Finger Style Ergonomic
For Use With (Application) Transporting a measured volume of liquid
For Use With (Equipment) D10, DL10, DF10ST, DFL10ST tips
Quantity 1
Width (English) 4.25 in
Fit for Purpose Certified Quality
Show More Show Less