Learn More
Gilson™ PIPETMANTM Multichannel Trade-In PROMO
GREENER_CHOICE
Send in your used or broken pipettes and save on select Gilson models. All used brands and models qualify and a pre-postage bag will be included with the order. Refurbished pipettes will be donated to Seeding Labs, who distributes to labs and universities around the world who need it most.
Supplier:  Gilson™ F144070T
 View more versions of this product
Catalog No. F144070TG
Only null left
Description
Description
  • PIPETMANTM Multichannel pipettes bring a new level of comfort into the Gilson range
  • Redesigned piston mechanism reduces the force needed to aspirate and dispense liquids
  • Offers one of the lowest pipetting forces commercially available
  • Maintains the trusted durability, accuracy, and reliability of the original PIPETMANTM pipettes
  • New piston assembly system and high quality piston grease guarantee hours of comfortable pipetting
Specifications
Chemical Identifiers
Diamond Tips
No
Manual
Ergonomic
Pipetman
Non-sterile
1
5.98 in
1.5 in
Adjustable Volume
Not Autoclavable
Reusable
Metal Ejector
No
Manual Air Displacement Pipette
3 Years
152 mm
38 mm
Specifications
Diamond Tips
No
Yellow
Reusable
Ergonomic
P8x20G
Pipetman
Non-sterile
2 to 20 μL
3 Years
330 mm
152 mm
38 mm
Adjustable Volume
Not Autoclavable
Manual
Pipette Tip 2 to 200 μL
Metal Ejector
8
No
Manual Air Displacement Pipette
1
330 x 152 x 38 mm
5.98 in
1.5 in
.81 lb
Product Suggestions
Product Suggestions