missing translation for 'onlineSavingsMsg'
Learn More

Gilson™ PIPETMAN™ Classic Pipette Trade-In PROMO

Catalog No. F144802TG
Change view
Click to view available options
Volume (Metric):
1 to 10 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
F144802TG 1 to 10 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F144802TG Supplier Gilson™ Supplier No. F144802T

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Send in your used or broken pipettes and save on select Gilson models. All used brands and models qualify and a pre-postage bag will be included with the order. Refurbished pipettes will be donated to Seeding Labs, who distributes to labs and universities around the world who need it most.

  • Fully adjustable, air-displacement pipette with selected volume range
  • Volume shown on digital indicator (volumeter)
  • Autoclavable tip holder and tip ejector help to avoid cross-contamination
  • Each pipette bears engraved identification number for traceability

Specifications

Accuracy ±0.025, ±2.5 μL
Description Only Tip Ejector, and Tip Holder are Autoclavable
Adjustable Tip Spacing No
Autoclavable Autoclavable
Color Red
Dispensing Speed Manual
Disposable Reusable
For Use With D-10, DL10, DF10ST, DFL10ST Tip
Grip Type Ergonomic
Material PVDF, Stainless Steel
Model P10
Number of Channels 1
Product Line Pipetman Classic
Programmability No
Sterility Non-sterile
Type Pipette
Volume (Metric) 1 to 10 μL
For Use With (Application) PCR, DNA sequencing
For Use With (Equipment) D10, DL10, DF10ST, DFL10ST tips
Fit for Purpose Certified Quality
Show More Show Less