missing translation for 'onlineSavingsMsg'
Learn More

Piperonyl Alcohol 98.0+%, TCI America™

Catalog No. P045725G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P045725G 25 g
P0457100G 100 g
P0457500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P045725G Supplier TCI America Supplier No. P045725G

Chemical Identifiers

CAS 495-76-1
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.149
MDL Number MFCD00005836
InChI Key BHUIUXNAPJIDOG-UHFFFAOYSA-N
Synonym piperonyl alcohol, piperonol, 1,3-benzodioxole-5-methanol, 3,4-methylenedioxybenzyl alcohol, benzo d 1,3 dioxol-5-ylmethanol, piperonylalcohol, 5-hydroxymethyl-1,3-benzodioxole, 1-hydroxymethyl-3,4-methylenedioxybenzene, benzo 1,3 dioxol-5-ylmethanol, 3,4-methylenedioxy phenylmethanol
PubChem CID 10322
IUPAC Name 1,3-benzodioxol-5-ylmethanol
SMILES C1OC2=C(O1)C=C(C=C2)CO

Specifications

Melting Point 54°C
Color White-Yellow
Boiling Point 157°C
Quantity 25 g
Formula Weight 152.15
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Piperonyl Alcohol
EINECSNumber (9)-1113
TSCA Yes