accessibility menu, dialog, popup

UserName

CAS RN 495-76-1

IUPAC Name: (2H-1,3-benzodioxol-5-yl)methanol
Synonyms: 1,3-benzodioxole-5-methanol
Molecular Weight (g/mol): 152.15
Molecular Formula: C8H8O3
InChi Key: BHUIUXNAPJIDOG-UHFFFAOYSA-N
SMILES: OCC1=CC=C2OCOC2=C1
Boiling Point 
  • (2)
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (5)
  • (2)
Percent Purity 
  • (3)
  • (4)
Quantity 
  • (3)
  • (3)
  • (1)
Physical Form 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (4)
  • (2)
  • (2)
  • (3)
  • (5)
  • (2)
  • (3)
  • (4)
  • (3)
  • (3)
  • (1)
  • (3)
  • (2)