missing translation for 'onlineSavingsMsg'
Learn More

Piperonyl alcohol, 98%

CAS: 495-76-1 | C8H8O3 | 152.15 g/mol

$259.24 - $259.24

Chemical Identifiers

CAS 495-76-1
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.15
MDL Number MFCD00005836
InChI Key BHUIUXNAPJIDOG-UHFFFAOYSA-N
Synonym piperonyl alcohol, piperonol, 1,3-benzodioxole-5-methanol, 3,4-methylenedioxybenzyl alcohol, benzo d 1,3 dioxol-5-ylmethanol, piperonylalcohol, 5-hydroxymethyl-1,3-benzodioxole, 1-hydroxymethyl-3,4-methylenedioxybenzene, benzo 1,3 dioxol-5-ylmethanol, 3,4-methylenedioxy phenylmethanol
PubChem CID 10322
IUPAC Name 1,3-benzodioxol-5-ylmethanol
SMILES C1OC2=C(O1)C=C(C=C2)CO
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC131351000
SDS View Documents
Thermo Scientific Chemicals
131351000
100 g Glass bottle
Each for $259.24
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

495-76-1
152.15
BHUIUXNAPJIDOG-UHFFFAOYSA-N
10322
C1OC2=C(O1)C=C(C=C2)CO
C8H8O3
MFCD00005836
piperonyl alcohol, piperonol, 1,3-benzodioxole-5-methanol, 3,4-methylenedioxybenzyl alcohol, benzo d 1,3 dioxol-5-ylmethanol, piperonylalcohol, 5-hydroxymethyl-1,3-benzodioxole, 1-hydroxymethyl-3,4-methylenedioxybenzene, benzo 1,3 dioxol-5-ylmethanol, 3,4-methylenedioxy phenylmethanol
1,3-benzodioxol-5-ylmethanol

Specifications

495-76-1
White
>110°C
97.5% min. (GC)
C8H8O3
100 g
piperonyl alcohol, piperonol, 1,3-benzodioxole-5-methanol, 3,4-methylenedioxybenzyl alcohol, benzo d 1,3 dioxol-5-ylmethanol, piperonylalcohol, 5-hydroxymethyl-1,3-benzodioxole, 1-hydroxymethyl-3,4-methylenedioxybenzene, benzo 1,3 dioxol-5-ylmethanol, 3,4-methylenedioxy phenylmethanol
BHUIUXNAPJIDOG-UHFFFAOYSA-N
1,3-benzodioxol-5-ylmethanol
10322
98%
Piperonyl alcohol
51.0°C to 55.0°C
156.0°C to 158.0°C (66.0 mmHg)
Authentic
Glass bottle
MFCD00005836
19, 67
Solubility in water: soluble. Other solubilities: soluble in ethanol, diethyl ether, benzene,, soluble in chloroform
C1OC2=C(O1)C=C(C=C2)CO
152.15
152.15
Crystalline Powder or Crystals
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 207-808-4

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only