missing translation for 'onlineSavingsMsg'
Learn More

Piperidine-1-carbonitrile 98.0+%, TCI America™

Catalog No. P23431G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P23431G 1 g
P23435G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P23431G Supplier TCI America Supplier No. P23431G
Only null left

Chemical Identifiers

CAS 1530-87-6
Molecular Formula C6H10N2
Molecular Weight (g/mol) 110.16
MDL Number MFCD00006477
InChI Key NVPICXQHSYQKGM-UHFFFAOYSA-N
Synonym 1-piperidinecarbonitrile, 1-cyanopiperidine, piperidinocarbonitrile, n-cyanopiperidine, pentamethylenecyanamide, cyanopiperidine, cyano-piperidine, piperidine nitrile, piperidinecarbonitrile, 1-cyanopiperidine #
PubChem CID 73736
IUPAC Name piperidine-1-carbonitrile
SMILES N#CN1CCCCC1

Specifications

Color Yellow
Boiling Point 103°C
Quantity 1 g
UN Number 3276
Formula Weight 110.16
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Piperidine-1-carbonitrile