missing translation for 'onlineSavingsMsg'
Learn More

Piperazine Phosphate Monohydrate 98.0+%, TCI America™

Catalog No. P049225G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P049225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P049225G Supplier TCI America Supplier No. P049225G
Only null left

Chemical Identifiers

CAS 14538-56-8
Molecular Formula C4H13N2O4P
Molecular Weight (g/mol) 184.132
MDL Number MFCD00036382
InChI Key NQQWFVUVBGSGQN-UHFFFAOYSA-N
PubChem CID 64733
IUPAC Name phosphoric acid;piperazine
SMILES C1CNCCN1.OP(=O)(O)O

Specifications

Color White
Quantity 25 g
Formula Weight 184.13
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Piperazine Phosphate Monohydrate