accessibility menu, dialog, popup

UserName

OHPOHOHONHHN

CAS RN 14538-56-8

IUPAC Name: phosphoric acid; piperazine
Synonyms: phosphoric acid; piperazine
Molecular Weight (g/mol): 184.13
Molecular Formula: C4H13N2O4P
InChi Key: NQQWFVUVBGSGQN-UHFFFAOYSA-N
SMILES: OP(O)(O)=O.C1CNCCN1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results