missing translation for 'onlineSavingsMsg'
Learn More

Piperazine hexahydrate, ≥99% (TLC), MilliporeSigma™ Supelco™

Catalog No. 111019727
Change view
Click to view available options
Quantity:
200 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019727 200 g
111019092 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111019727 Supplier Merck Emd Millipore Supplier No. P7003200G

Piperazine hexahydrate may be used as a standard for the determination of piperazine in pharmaceutical preparations using colorimetric technique.

Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 142-63-2
Vapor Pressure 0.8 mmHg (20°C)
Flash Point 88°C
Physical Form Neat
Linear Formula C4H10N2 · 6H2 O
MDL Number MFCD00149389
Melting Point 42°C to 44°C
Boiling Point 145°C to 156°C
Type Analytical Standard
Quantity 200 g
Formula Weight 194.23
Percent Purity ≥99% (TLC)
Grade Analytical Standard
Show More Show Less