missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Piperazine Dihydriodide 98.0+%, TCI America™
Supplier: TCI America P24921G
Specifications
| Piperazine Dihydriodide | |
| 250°C | |
| MFCD00054377 | |
| Piperazinium Diiodide, Piperazinium Iodide | |
| I.I.C1CNCCN1 | |
| 341.96 | |
| 341.96 | |
| Crystalline Powder |
| 58464-47-4 | |
| C4H12I2N2 | |
| 1 g | |
| QZCGFUVVXNFSLE-UHFFFAOYSA-N | |
| piperazine dihydroiodide | |
| 20068687 | |
| ≥98.0% (N,T) |
Chemical Identifiers
| 58464-47-4 | |
| 341.96 | |
| QZCGFUVVXNFSLE-UHFFFAOYSA-N | |
| 20068687 | |
| I.I.C1CNCCN1 |
| C4H12I2N2 | |
| MFCD00054377 | |
| Piperazinium Diiodide, Piperazinium Iodide | |
| piperazine dihydroiodide |
Safety and Handling
TSCA : No