accessibility menu, dialog, popup

UserName

CAS RN 58464-47-4

NHHNHIHI

CAS RN 58464-47-4

IUPAC Name: piperazine dihydroiodide
Synonyms: piperazine dihydroiodide
Molecular Weight (g/mol): 341.96
Molecular Formula: C4H12I2N2
InChi Key: QZCGFUVVXNFSLE-UHFFFAOYSA-N
SMILES: I.I.C1CNCCN1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results