missing translation for 'onlineSavingsMsg'
Learn More

Piperazine Anhydrous 98.0+%, TCI America™

Catalog No. P044625G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P044625G 25 g
P0446500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P044625G Supplier TCI America Supplier No. P044625G

Chemical Identifiers

CAS 110-85-0
Molecular Formula C4H10N2
Molecular Weight (g/mol) 86.14
MDL Number MFCD00005953
InChI Key GLUUGHFHXGJENI-UHFFFAOYSA-N
Synonym diethylenediamine, piperazin, 1,4-diazacyclohexane, hexahydropyrazine, piperazidine, antiren, 1,4-piperazine, diethyleneimine, eraverm, pipersol
PubChem CID 4837
ChEBI CHEBI:28568
IUPAC Name piperazine
SMILES C1CNCCN1

Specifications

Melting Point 112°C
Color White
pH 10.8 to 11.8 (10% soln.H2O)
Boiling Point 146°C
Quantity 25 g
UN Number 2579
Formula Weight 86.14
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Piperazine Anhydrous
EINECSNumber (5)-0953
RTECSNumber TK7800000
TSCA Yes