accessibility menu, dialog, popup

UserName

CAS RN 110-85-0

IUPAC Name: piperazine
Synonyms: piperazine piperazine phosphate (substance) wurmirazin valamina diethyleneimine lyogen anatensol vermex pyrazine hexahydride pacinol
Molecular Weight (g/mol): 86.14
Molecular Formula: C4H10N2
InChi Key: GLUUGHFHXGJENI-UHFFFAOYSA-N
SMILES: C1CNCCN1
Quantity 
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
  • (1)
Boiling Point 
  • (2)
  • (4)
  • (3)
  • (7)
Percent Purity 
  • (2)
  • (7)
  • (7)
Physical Form 
  • (3)
  • (2)
  • (7)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (14)
  • (1)
  • (7)
  • (2)
  • (7)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
  • (1)
  • (2)
  • (4)
  • (3)
  • (7)
  • (2)
  • (7)
  • (7)
  • (3)
  • (2)
  • (7)
  • (3)
  • (7)