missing translation for 'onlineSavingsMsg'
Learn More

Piperazine, anhydrous, 99%

Catalog No. p-7022328
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
5 x 500 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1504922 100 g
AAA1504936 500 g
AAA15049A1 1 kg
AAA1504961 5 x 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AAA1504922 Supplier Thermo Scientific Chemicals Supplier No. A1504922
Only null left

CAS: 110-85-0 | C4H10N2 | 86.14 g/mol

Piperazine acts as an intermediate for pharmaceuticals, polymers, dyes, corrosion inhibitors, rubber accelerators and surfactants. It is used as a raw material in the manufacture of plastics, polyamide resins, urethane and brake fluid. It is also used for carbondioxide and hydrogen sulfide gas scrubbing mediums. Furthermore, it serves as an epoxy curing agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Piperazine acts as an intermediate for pharmaceuticals, polymers, dyes, corrosion inhibitors, rubber accelerators and surfactants. It is used as a raw material in the manufacture of plastics, polyamide resins, urethane and brake fluid. It is also used for carbondioxide and hydrogen sulfide gas scrubbing mediums. Furthermore, it serves as an epoxy curing agent.

Solubility
Soluble in water, chloroform, glycerol, glycols and alcohol. Slightly soluble in benzene and heptane. Insoluble in ether.

Notes
Air and light sensitive. Hygroscopic. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents and strong acids.

Chemical Identifiers

CAS 110-85-0
Molecular Formula C4H10N2
Molecular Weight (g/mol) 86.14
MDL Number MFCD00005953
InChI Key GLUUGHFHXGJENI-UHFFFAOYSA-N
Synonym diethylenediamine, piperazin, 1,4-diazacyclohexane, hexahydropyrazine, piperazidine, antiren, 1,4-piperazine, diethyleneimine, eraverm, pipersol
PubChem CID 4837
ChEBI CHEBI:28568
IUPAC Name piperazine
SMILES C1CNCCN1

Specifications

Melting Point 108°C to 113°C
Density 1.11
Boiling Point 146°C to 148°C
Flash Point 88°C (190°F)
Quantity 100 g
UN Number UN2579
Beilstein 102555
Sensitivity Hygroscopic
Merck Index 14,7464
Solubility Information Soluble in water,chloroform,glycerol,glycols and alcohol. Slightly soluble in benzene and heptane. Insoluble in ether.
Formula Weight 86.14
Percent Purity 99%
Chemical Name or Material Piperazine, anhydrous
Show More Show Less
Hazard Category H228-H314-H317-H334-H361fd
Hazard Statement GHS H Statement
H334-H361-H314-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Suspected of damaging fertility or the unborn child.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P210-P240-P241-P260-P264b-P272-P280g-P281-P285-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P333+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: PIPERAZINE
EINECSNumber 203-808-3
RTECSNumber TK7800000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only