missing translation for 'onlineSavingsMsg'
Learn More

Piperazine, Anhydrous, 99%, Spectrum™ Chemical

Catalog No. p-7166213
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
12 kg
25 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18612385 5 g Amber Glass Bottle
18612381 100 g Amber Glass Bottle
18612387 500 g Amber Glass Bottle
18612383 2.5 kg Poly Pail
18612382 12 kg Poly Pail
18612384 25 kg Fiber Drum
18612386 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 18612385 Supplier Spectrum Chemical Mfg Cor Supplier No. PI1235GM
Only null left

Chemical Identifiers

CAS 110-85-0
Molecular Formula C4H10N2
Molecular Weight (g/mol) 86.14
InChI Key GLUUGHFHXGJENI-UHFFFAOYSA-N
IUPAC Name piperazine
SMILES C1CNCCN1

Specifications

CAS Max % 1
Melting Point 108° to 113°C
Packaging Amber Glass Bottle
Quantity 5 g
Percent Purity 0.99
Grade Reagent

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.