missing translation for 'onlineSavingsMsg'
Learn More

Piperazine-1,4-bis(2-hydroxypropanesulfonic Acid) Dihydrate, TCI America™

Catalog No. p113125g
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P113125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P113125G Supplier TCI America Supplier No. P113125G

[Good's buffer component for biological research]

Chemical Identifiers

CAS 68189-43-5
Molecular Formula C10H22N2O8S2
Molecular Weight (g/mol) 362.412
MDL Number MFCD00038350
InChI Key LVQFQZZGTZFUNF-UHFFFAOYSA-N
Synonym popso, piperazine-1,4-bis 2-hydroxypropanesulfonic acid, piperazine-n,n'-bis 2-hydroxypropanesulfonic acid, piperazine-1,4-bis 2-hydroxypropanesulfonic acid dihydrate, popso hydrate, 3,3'-piperazine-1,4-diyl bis 2-hydroxypropane-1-sulfonic acid, 2-hydroxy-3-4-2-hydroxy-3-sulfopropyl piperazin-1-yl propane-1-sulfonic acid, 2-hydroxy-3-4-2-hydroxy-3-sulfopropyl piperazinyl propanesulfonic acid, 1,4-piperazinedipropanesulfonic acid,, a,, a'-dihydroxy, beta,beta'-dihydroxypiperazine-1,4-dipropanesulphonic acid
PubChem CID 109211
IUPAC Name 2-hydroxy-3-[4-(2-hydroxy-3-sulfopropyl)piperazin-1-yl]propane-1-sulfonic acid
SMILES C1CN(CCN1CC(CS(=O)(=O)O)O)CC(CS(=O)(=O)O)O

Specifications

Color White
Quantity 25 g
Formula Weight 362.41
Physical Form Crystalline Powder
Chemical Name or Material Piperazine-1,4-bis(2-hydroxypropanesulfonic Acid) Dihydrate [Good′s buffer component for biolo