accessibility menu, dialog, popup

UserName

HO(S)NHNH(S)HOSOOOSOOO

CAS RN 68189-43-5

IUPAC Name: 1,4-bis[(2S)-2-hydroxy-3-sulfonatopropyl]piperazine-1,4-diium
Synonyms: 1,4-bis[(2S)-2-hydroxy-3-sulfonatopropyl]piperazine-1,4-diium
Molecular Weight (g/mol): 362.41
Molecular Formula: C10H22N2O8S2
InChi Key: LVQFQZZGTZFUNF-UWVGGRQHSA-N
SMILES: O[C@@H](C[NH+]1CC[NH+](C[C@H](O)CS([O-])(=O)=O)CC1)CS([O-])(=O)=O
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results