missing translation for 'onlineSavingsMsg'
Learn More

Piperacillin Sodium Salt 98.0+%, TCI America™

Catalog No. p-7135247
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P17741G 1 g
P17745G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P17741G Supplier TCI America Supplier No. P17741G
Only null left

Chemical Identifiers

CAS 59703-84-3
Molecular Formula C23H26N5NaO7S
Molecular Weight (g/mol) 539.54
MDL Number MFCD00917471
InChI Key WCMIIGXFCMNQDS-QMJSIVKPSA-M
Synonym dsstox_cid_28891, dsstox_rid_83159, dsstox_gsid_48965, piperacillin sodium salt
PubChem CID 23666879
ChEBI CHEBI:8233
IUPAC Name sodium (2S,5R,6R)-6-[(2S)-2-[(4-ethyl-2,3-dioxopiperazine-1-carbonyl)amino]-2-phenylacetamido]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
SMILES [Na+].CCN1CCN(C(=O)N[C@H](C(=O)N[C@H]2[C@H]3SC(C)(C)[C@@H](N3C2=O)C([O-])=O)C2=CC=CC=C2)C(=O)C1=O

Specifications

Melting Point 185°C
Color White
Quantity 1 g
Formula Weight 539.54
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Piperacillin Sodium Salt
RTECSNumber XI0180000
TSCA No