missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Piperacillin Sodium Salt, MP Biomedicals™
Specifications
Specifications
| Type | Piperacillin Sodium Salt |
| Physical Form | Crystalline Powder |
| Color | White |
| Quantity | 10 g |
| IUPAC Name | sodium (2S,5R,6R)-6-[(2S)-2-[(4-ethyl-2,3-dioxopiperazine-1-carbonyl)amino]-2-phenylacetamido]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate |
| Formula Weight | 539.5 |
| Grade | Not Available |
| Chemical Name or Material | Piperacillin Sodium Salt |
Safety and Handling
| Recommended Storage | Store at 4°C. |